Assay Details
Target Gene Details
Entrez Gene ID: | 107817 |
Gene Name: | jumonji domain containing 6 |
Gene Aliases: |
5730436I23Rik, D11Ertd195e, PSR, PtdSerR, Ptdsr, mKIAA0585 |
Location: |
Chr.11:116837432-116843449 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jmjd6 | NM_033398.2 | 2 | 709 | NP_203971.2 |
| AF304118.1 | 2 | 642 | AAG27719.1 | |
| AK122317.1 | 2 | 550 | BAC65599.1 | |
| BC056629.1 | 2 | 709 | AAH56629.1 | |
| BC117737.1 | 1 | 350 | ||
| CX210791.1 | 2 | 415 | ||
| EF527404.1 | 2 | 709 | ||
| EF527405.1 | 2 | 495 |
More Information
Set Membership: |
|

Genomic Map