Assay Details
Target Gene Details
Entrez Gene ID: | 16480 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
Ctnng, D930025P04Rik, PG |
Location: |
Chr.11:100368856-100397790 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jup | NM_010593.2 | 14 | 3105 | NP_034723.1 |
| AK086401.1 | 2 | 1116 | ||
| AK145895.1 | 14 | 3090 | ||
| AK170934.1 | 14 | 3078 | BAE42126.1 | |
| BC040757.1 | 14 | 3054 | AAH40757.1 |

Genomic Map