Assay Details
Target Gene Details
Entrez Gene ID: | 21944 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 12 |
Gene Aliases: |
Apo3l, Dr3l, Dr3lg, Tweak |
Location: |
Chr.11:69686240-69696098 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf12 | NM_011614.3 | 7 | 1090 | NP_035744.1 |
| AF030100.2 | 7 | 746 | AAC53517.2 | |
| AK020909.1 | 4 | 429 | BAB32249.1 | |
| AK170297.1 | 7 | 990 | BAE41696.1 | |
| AK171014.1 | 7 | 990 | BAE42185.1 | |
| BX634398.1 | ||||
| CB204861.1 | 2 | 368 |
Target Gene Details
Entrez Gene ID: | 619441 |
Gene Name: | tumor necrosis factor (ligand) superfamily, membrane-bound member 13 |
Gene Aliases: |
Tnfsf12-Tnfsf13, Tnfsf12Tnfsf13, Tnfsf12tnfsf13, Twe-pril |
Location: |
Chr.11:69682577-69696098 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsfm13 | NM_001034097.2 | NP_001029269.1 | ||
| NM_001159503.2 | NP_001152975.1 | |||
| AK044387.1 | BAC31897.1 | |||
| BC096441.1 | AAH96441.1 |
More Information
Set Membership: |
|

Genomic Map