Assay Details
Target Gene Details
Entrez Gene ID: | 69583 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 13 |
Gene Aliases: |
2310026N09Rik, April, Tall2, Trdl1 |
Location: |
Chr.11:69682577-69685554 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf13 | NM_001159505.1 | 6 | 1421 | NP_001152977.1 |
| NM_023517.2 | 6 | 1424 | NP_076006.2 | |
| AK040667.1 | BAC30661.1 | |||
| AK134028.1 | 6 | 1127 | BAE21985.1 | |
| BY463961.1 | 1 | 191 |
Target Gene Details
Entrez Gene ID: | 619441 |
Gene Name: | tumor necrosis factor (ligand) superfamily, membrane-bound member 13 |
Gene Aliases: |
Tnfsf12-Tnfsf13, Tnfsf12Tnfsf13, Tnfsf12tnfsf13, Twe-pril |
Location: |
Chr.11:69682577-69696098 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsfm13 | NM_001034097.2 | 13 | 1980 | NP_001029269.1 |
| NM_001159503.2 | 13 | 1974 | NP_001152975.1 | |
| AK044387.1 | 13 | 1981 | BAC31897.1 |

Genomic Map