Assay Details
Target Gene Details
Entrez Gene ID: | 104111 |
Gene Name: | adenylate cyclase 3 |
Gene Aliases: |
AC3, mKIAA0511 |
Location: |
Chr.12:4133087-4213525 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adcy3 | NM_001159536.1 | 1 | 260 | NP_001153008.1 |
NM_001159537.1 | 1 | 260 | NP_001153009.1 | |
AK122298.1 | 1 | 231 | BAC65580.1 | |
BC057113.1 | 1 | 189 | AAH57113.1 | |
BC057316.1 | 1 | 84 | AAH57316.1 | |
CF532558.1 | 1 | 553 |
