Assay Details
Target Gene Details
Entrez Gene ID: | 268379 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 13 |
Gene Aliases: |
9830132L24, A930002G16Rik, AI956815 |
Location: |
Chr.11:9191354-9684871 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca13 | NM_178259.3 | 18 | 7747 | NP_839990.2 |
More Information
Set Membership: |
|

Genomic Map