Assay Details
Target Gene Details
Entrez Gene ID: | 217306 |
Gene Name: | CD300e antigen |
Gene Aliases: |
BC034097, CLM2, Cd300le, TREM5 |
Location: |
Chr.11:115051917-115075950 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300e | NM_172050.2 | 4 | 1468 | NP_742047.1 |
| BC034097.1 | 4 | 1493 | AAH34097.1 | |
| BC039971.1 | 4 | 1504 | AAH39971.1 |
More Information
Set Membership: |
|

Genomic Map