Assay Details
Target Gene Details
Entrez Gene ID: | 56494 |
Gene Name: | golgi SNAP receptor complex member 2 |
Gene Aliases: |
2310032N09Rik, C76855, Gs27, SNARE, membrin |
Location: |
Chr.11:103676849-103697738 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gosr2 | NM_019650.3 | 6 | 956 | NP_062624.2 |
| AK013052.1 | 6 | 950 | BAB28622.1 | |
| AK150004.1 | 6 | 954 | BAE29231.1 | |
| BC008525.1 | 6 | 952 | AAH08525.1 | |
| BC051253.1 | 6 | 922 | AAH51253.1 |
More Information
Set Membership: |
|

Genomic Map