Assay Details
Target Gene Details
Entrez Gene ID: | 214384 |
Gene Name: | myocardin |
Gene Aliases: |
BSAC2A, Srfcp |
Location: |
Chr.11:65173608-65269989 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myocd | NM_145136.4 | 8 | 1053 | NP_660118.3 |
| NM_146386.3 | 8 | 1053 | NP_666498.2 | |
| AF384055.2 | 8 | 1019 | AAK71683.2 | |
| AF437877.1 | 4 | 378 | AAL30892.1 | |
| AK142216.1 | 8 | 1056 | BAE24980.1 | |
| AY303755.1 | 8 | 1044 | AAQ63841.1 | |
| BC141281.1 | 9 | 1099 |
More Information
Set Membership: |
|

Genomic Map