Assay Details
Target Gene Details
Entrez Gene ID: | 58172 |
Gene Name: | SERTA domain containing 2 |
Gene Aliases: |
AB041541, AU021878, Kiaa0127, MNCb-1504, SEI-2, Sei2, TRIP-Br2, mKIAA0127 |
Location: |
Chr.11:20542938-20653023 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sertad2 | NM_001038625.1 | 1 | 96 | NP_001033714.1 |
| NM_021372.2 | NP_067347.2 | |||
| AB041541.1 | BAA95026.1 | |||
| AK132846.1 | 1 | 76 | BAE21389.1 | |
| AK142018.1 | 1 | 76 | BAE24915.1 | |
| AK146621.1 | 1 | 97 | BAE27310.1 | |
| AK146897.1 | 1 | 97 | ||
| AK147674.1 | BAE28066.1 | |||
| BC014726.1 | AAH14726.1 |
More Information
Set Membership: |
|

Genomic Map