Assay Details
Target Gene Details
Entrez Gene ID: | 58172 |
Gene Name: | SERTA domain containing 2 |
Gene Aliases: |
AB041541, AU021878, Kiaa0127, MNCb-1504, SEI-2, Sei2, TRIP-Br2, mKIAA0127 |
Location: |
Chr.11:20542938-20653023 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sertad2 | NM_021372.2 | 1 | 254 | NP_067347.2 |
| AB041541.1 | 1 | 173 | BAA95026.1 | |
| AK142268.1 | 1 | 255 | ||
| AK147674.1 | 1 | 345 | BAE28066.1 | |
| BC014726.1 | 1 | 87 | AAH14726.1 |
More Information
Set Membership: |
|

Genomic Map