Assay Details
Target Gene Details
Entrez Gene ID: | 22057 |
Gene Name: | transducer of ErbB-2.1 |
Gene Aliases: |
Tob, Trob |
Location: |
Chr.11:94211454-94215495 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tob1 | NM_009427.2 | 2 | 609 | NP_033453.2 |
| AK142667.1 | 2 | 625 | BAE25152.1 | |
| BC082588.1 | 2 | 609 | AAH82588.1 | |
| CF748516.1 |

Genomic Map