Assay Details
Target Gene Details
Entrez Gene ID: | 217258 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 8a |
Gene Aliases: |
- |
Location: |
Chr.11:110025634-110097583 on Build GRCm38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca8a | NM_153145.4 | 39 | 4970 | NP_694785.3 |
| AB255881.1 | ||||
| AB255882.1 | ||||
| AF498360.1 | 38 | 4898 | AAM90906.1 | |
| AK031843.1 | 20 | 2201 | BAC27576.1 | |
| AY732492.1 | 38 | 4898 | AAU81985.1 | |
| BC026496.1 | 19 | 2095 | AAH26496.1 | |
| BC060032.1 | 21 | 2464 | AAH60032.1 |
More Information
Set Membership: |
|

Genomic Map