Assay Details
Target Gene Details
Entrez Gene ID: | 192197 |
Gene Name: | breast carcinoma amplified sequence 3 |
Gene Aliases: |
1500019F07Rik, 2610028P08Rik, AU021018, BC028339, K20D4, rudhira |
Location: |
Chr.11:85353116-85826058 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcas3 | NM_001166642.1 | 16 | 1555 | NP_001160114.1 |
NM_001166643.1 | 15 | 1469 | NP_001160115.1 | |
NM_138681.4 | 16 | 1555 | NP_619622.3 | |
AF313800.1 | 15 | 1449 | AAG34697.1 | |
AK032423.1 | 16 | 1506 | BAC27862.1 | |
AK034117.1 | 16 | 1535 | BAC28592.1 | |
AK157222.1 | 16 | 1505 | ||
AK160127.1 | 15 | 1448 | ||
AK171964.1 | 15 | 1422 | BAE42749.1 | |
AK192083.1 | 1 | 105 | ||
BC118965.2 | 15 | 1469 | ||
BC118966.1 | 15 | 1451 |
