Assay Details
Target Gene Details
Entrez Gene ID: | 70357 |
Gene Name: | Kv channel-interacting protein 1 |
Gene Aliases: |
KCHIP1, Kchip1.2 |
Location: |
Chr.11:33629339-33993193 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnip1 | NM_001190885.1 | 1 | 323 | NP_001177814.1 |
| NM_001190886.1 | NP_001177815.1 | |||
| NM_001290690.1 | 1 | 323 | NP_001277619.1 | |
| NM_027398.3 | 1 | 323 | NP_081674.2 | |
| AK013657.1 | 1 | 301 | ||
| AK014313.1 | 1 | 312 | ||
| AK045948.1 | 1 | 324 | BAC32543.1 | |
| AK046398.1 | 1 | 312 | BAC32705.1 | |
| AK048178.1 | ||||
| AK081845.1 | BAC38347.1 | |||
| AK082267.1 | ||||
| AY171234.1 | 1 | 199 | AAN77492.1 | |
| AY647242.1 | 1 | 155 | AAT68468.1 | |
| BC034241.1 | 1 | 265 | AAH34241.1 | |
| DQ148493.1 | 1 | 265 | AAZ77810.1 | |
| DQ148494.1 | AAZ77811.1 |

Genomic Map