Assay Details
Target Gene Details
Entrez Gene ID: | 110809 |
Gene Name: | serine/arginine-rich splicing factor 1 |
Gene Aliases: |
1110054N12Rik, 5730507C05Rik, 6330415C05Rik, AI482334, AW491331, Asf, Sf2, Sfrs1 |
Location: |
Chr.11:88046902-88053757 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf1 | NM_001078167.2 | 3 | 4105 | NP_001071635.1 |
| NM_173374.4 | 4 | 3909 | NP_775550.2 | |
| AK017754.1 | 1 | 605 |
More Information
Set Membership: |
|

Genomic Map