Assay Details
Target Gene Details
Entrez Gene ID: | 20382 |
Gene Name: | serine/arginine-rich splicing factor 2 |
Gene Aliases: |
D11Wsu175e, MRF-1, Pr264, SC35, Sfrs10, Sfrs2 |
Location: |
Chr.11:116849897-116853104 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf2 | NM_011358.2 | 3 | 1547 | NP_035488.1 |
| AF077858.1 | 3 | 1535 | AAC71000.1 | |
| AF250132.1 | 2 | 820 | AAK28416.1 | |
| AF250133.1 | 1 | 994 | ||
| AF250134.1 | 1 | 470 | ||
| AF250135.1 | 2 | 1458 | AAK28417.1 | |
| AK076448.1 | 3 | 1538 | BAC36346.1 | |
| AK086103.1 | 3 | 1545 | BAC39610.1 | |
| AK088042.1 | 3 | 1545 | BAC40111.1 | |
| AK161873.1 | 3 | 1543 | ||
| AK165670.1 | 3 | 1549 | BAE38330.1 | |
| AK177537.1 | 2 | 195 | ||
| AK207032.1 | 1 | 310 | ||
| AK211203.1 | 1 | 306 | ||
| AK212169.1 | 1 | 321 | ||
| AK220014.1 | 1 | 324 | ||
| BC005493.1 | 5 | 1127 | AAH05493.1 | |
| BG093253.1 | ||||
| L13610.1 | 4 | 939 | AAA39332.1 | |
| U14648.1 | 3 | 1542 | AAA64595.1 |
More Information
Set Membership: |
|

Genomic Map