Assay Details
Target Gene Details
Entrez Gene ID: | 217337 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
2610024I03Rik |
Location: |
Chr.11:116245166-116274217 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srp68 | NM_146032.3 | 1 | 240 | NP_666144.3 |
| AK033010.1 | 1 | 159 | BAC28124.1 | |
| AK076003.1 | 1 | 139 | ||
| AK180890.1 | ||||
| AK212180.1 | ||||
| AK212446.1 | ||||
| AK213152.1 | ||||
| BC070422.1 | 1 | 240 | AAH70422.1 |
More Information
Set Membership: |
|

Genomic Map