Assay Details
Target Gene Details
Entrez Gene ID: | 217337 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
2610024I03Rik |
Location: |
Chr.11:116245166-116274217 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srp68 | NM_146032.3 | 16 | 1888 | NP_666144.3 |
| AK033010.1 | 16 | 1807 | BAC28124.1 | |
| AK076003.1 | 16 | 1785 | ||
| AK210149.1 | 2 | 193 | ||
| AK211431.1 | ||||
| BC033573.1 | 12 | 1188 | AAH33573.1 | |
| BC070422.1 | 16 | 1888 | AAH70422.1 |
More Information
Set Membership: |
|

Genomic Map