Assay Details
Target Gene Details
Entrez Gene ID: | 69806 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 11 |
Gene Aliases: |
1810074D23Rik |
Location: |
Chr.11:113244851-113650752 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a11 | AK165545.1 |
Target Gene Details
Entrez Gene ID: | 20606 |
Gene Name: | somatostatin receptor 2 |
Gene Aliases: |
SSTR-2, Smstr-2, Smstr2, sst2 |
Location: |
Chr.11:113619481-113625895 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sstr2 | NM_001042606.2 | 2 | 1062 | NP_001036071.1 |
| NM_009217.4 | 2 | 1062 | NP_033243.2 | |
| S71756.1 | 1 | 537 | AAP20804.1 |
More Information
Set Membership: |
|

Genomic Map