Assay Details
Target Gene Details
Entrez Gene ID: | 217069 |
Gene Name: | tripartite motif-containing 25 |
Gene Aliases: |
AA960166, AL022677, EFP, Zfp147 |
Location: |
Chr.11:88999403-89020293 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim25 | NM_009546.2 | 1 | 115 | NP_033572.2 |
AK154533.1 | 1 | 108 | BAE32658.1 | |
AK160898.1 | 1 | 67 | BAE36077.1 | |
AK169562.1 | 1 | 116 | BAE41230.1 | |
AK172555.1 | 1 | 55 | ||
BC006908.1 | 1 | 84 | AAH06908.1 | |
BC034276.1 | 1 | 87 | AAH34276.1 | |
D63902.1 | 1 | 38 | BAA09941.1 |
