Assay Details
Target Gene Details
Entrez Gene ID: | 27225 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 |
Gene Aliases: |
1700055J08Rik, 2510027P10Rik, AI649272 |
Location: |
Chr.12:103407976-103425867 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx24 | NM_001159502.1 | 1 | 128 | NP_001152974.1 |
NM_020494.3 | NP_065240.2 | |||
AF214732.1 | AAG02170.1 | |||
AK011012.1 | BAB27330.1 | |||
AK018862.1 | 1 | 131 | ||
AK036538.1 | ||||
AK134339.1 | BAE22104.1 | |||
AK149810.1 | ||||
AK150778.1 | BAE29843.1 | |||
AK152551.1 | BAE31305.1 | |||
AK152928.1 | BAE31603.1 | |||
AK166360.1 | BAE38731.1 | |||
AK172557.1 | ||||
BC055048.1 | AAH55048.1 | |||
BC055317.1 | 1 | 128 | AAH55317.1 | |
CJ141277.1 |
