Assay Details
Target Gene Details
Entrez Gene ID: | 27225 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 |
Gene Aliases: |
1700055J08Rik, 2510027P10Rik, AI649272 |
Location: |
Chr.12:103407976-103425867 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx24 | NM_001159502.1 | 3 | 1074 | NP_001152974.1 |
NM_020494.3 | 3 | 908 | NP_065240.2 | |
AF214732.1 | 3 | 888 | AAG02170.1 | |
AK134339.1 | 3 | 886 | BAE22104.1 | |
AK149810.1 | 3 | 860 | ||
AK150778.1 | 3 | 860 | BAE29843.1 | |
AK152551.1 | 3 | 860 | BAE31305.1 | |
AK152928.1 | 3 | 860 | BAE31603.1 | |
AK166360.1 | 3 | 866 | BAE38731.1 | |
AK172557.1 | 3 | 860 | ||
BC055048.1 | 3 | 838 | AAH55048.1 | |
BC055317.1 | 3 | 1074 | AAH55317.1 |
