Assay Details
Target Gene Details
Entrez Gene ID: | 110417 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class H |
Gene Aliases: |
2210416H01Rik, A930028P05Rik |
Location: |
Chr.12:79080664-79089754 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigh | NM_029988.2 | 4 | 2042 | NP_084264.1 |
| AK167892.1 | 4 | 2024 | BAE39904.1 | |
| BC086806.1 | 4 | 1993 | AAH86806.1 |
Target Gene Details
Entrez Gene ID: | 211945 |
Gene Name: | pleckstrin homology domain containing, family H (with MyTH4 domain) member 1 |
Gene Aliases: |
AV308913, C77772, D630024D12Rik, mKIAA1200 |
Location: |
Chr.12:79029163-79081655 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhh1 | NM_181073.3 | 29 | 5764 | NP_851418.2 |
| AK052692.1 | 16 | 3439 | BAC35099.1 | |
| AK077737.1 | 1 | 1458 | ||
| AK122464.1 | 29 | 5733 | BAC65746.1 | |
| CF538481.1 | 1 | 299 |
More Information
Set Membership: |
|

Genomic Map