Assay Details
Target Gene Details
Entrez Gene ID: | 20716 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3N |
Gene Aliases: |
Spi2-2, Spi2.2, Spi2/eb.4 |
Location: |
Chr.12:104406675-104414334 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina3n | NM_009252.2 | 5 | 1314 | NP_033278.2 |
| BC013651.1 | 5 | 1293 | AAH13651.1 | |
| M64086.1 | 5 | 1269 | AAA40130.1 |
More Information
Set Membership: |
|

Genomic Map