Assay Details
Target Gene Details
Entrez Gene ID: | 268591 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 5 |
Gene Aliases: |
4933415L04, PAI-3, Pci |
Location: |
Chr.12:104099313-104106138 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina5 | NM_172953.3 | 2 | 568 | NP_766541.2 |
| AK077160.1 | 2 | 569 | BAC36651.1 | |
| AK087714.1 | 2 | 531 | BAC39979.1 | |
| BC090981.1 | 2 | 531 | AAH90981.1 |
More Information
Set Membership: |
|

Genomic Map