Assay Details
Target Gene Details
Entrez Gene ID: | 70059 |
Gene Name: | degenerative spermatocyte homolog 2 (Drosophila), lipid desaturase |
Gene Aliases: |
2210008A03Rik, AI852933, DES2 |
Location: |
Chr.12:108686792-108702306 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Degs2 | NM_001171002.1 | NP_001164473.1 | ||
| NM_027299.5 | 3 | 954 | NP_081575.2 | |
| AF466377.1 | 3 | 942 | AAM12533.1 | |
| AK008666.1 | 3 | 955 | ||
| AK162979.1 | BAE37145.1 | |||
| BC016427.1 | 3 | 936 | ||
| BU757432.1 | 1 | 343 |
More Information
Set Membership: |
|

Genomic Map