Assay Details
Target Gene Details
Entrez Gene ID: | 14874 |
Gene Name: | glutathione transferase zeta 1 (maleylacetoacetate isomerase) |
Gene Aliases: |
MAAI |
Location: |
Chr.12:87147165-87164723 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstz1 | NM_001252555.1 | 1 | 122 | NP_001239484.1 |
| AB041613.1 | 1 | 122 | BAA95096.1 |
Target Gene Details
Entrez Gene ID: | 217734 |
Gene Name: | protein-O-mannosyltransferase 2 |
Gene Aliases: |
A830009D15Rik, AW046274 |
Location: |
Chr.12:87106861-87147902 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pomt2 | NM_153415.3 | 1 | 617 | NP_700464.2 |
| AF246235.1 | 1 | 221 | AAL85627.1 | |
| AK043573.1 | 1 | 317 | ||
| AK043713.1 | 1 | 159 | ||
| AK050915.1 | 1 | 186 | BAC34458.1 | |
| AK076813.1 | 1 | 618 | ||
| AK085703.1 | 1 | 684 | ||
| AY090481.1 | 1 | 617 | AAM12047.1 | |
| AY090482.1 | 1 | 217 | AAM12048.1 | |
| BC052045.1 | 1 | 189 | AAH52045.1 | |
| BC062965.1 | 1 | 189 | AAH62965.1 |
More Information
Set Membership: |
|

Genomic Map