Assay Details
Target Gene Details
Entrez Gene ID: | 18193 |
Gene Name: | nuclear receptor-binding SET-domain protein 1 |
Gene Aliases: |
AI528500, KMT3B |
Location: |
Chr.13:55209782-55318325 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsd1 | NM_008739.3 | 18 | 5781 | NP_032765.3 |
CA318563.1 | 3 | 204 | ||
CN529078.1 |
