Assay Details
Target Gene Details
Entrez Gene ID: | 18193 |
Gene Name: | nuclear receptor-binding SET-domain protein 1 |
Gene Aliases: |
AI528500, KMT3B |
Location: |
Chr.13:55209782-55318325 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsd1 | NM_008739.3 | 2 | 558 | NP_032765.3 |
AK049663.1 | 2 | 514 | ||
AK053386.1 | ||||
AK135300.1 | ||||
AK145783.1 | 2 | 559 | BAE26648.1 | |
AK164087.1 | ||||
AK172676.1 |
