Assay Details
Target Gene Details
Entrez Gene ID: | 12050 |
Gene Name: | BCL2-like 2 |
Gene Aliases: |
AW048834, Bcl-w, Gtrgal2, Gtrosa41, bclw, c98 |
Location: |
Chr.14:54883366-54889095 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl2l2 | NM_007537.1 | 4 | 1558 | NP_031563.1 |
AF030769.1 | 4 | 1558 | AAB86430.1 | |
AK004680.1 | 4 | 1588 | BAB23468.1 | |
AK015644.1 | 4 | 1511 | BAB29912.1 | |
AK172925.1 | 2 | 1385 | BAD32203.1 | |
AY170344.3 | 4 | 1539 | AAO13177.2 | |
BC040369.1 | 2 | 2049 | AAH40369.1 |
