Assay Details
Target Gene Details
Entrez Gene ID: | 544971 |
Gene Name: | B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
Gene Aliases: |
AI662220, AW049390, AW456745, B130055N23Rik, G630013P12Rik, TAF3B1, TFC5, TFIIIB150, TFIIIB90, Tfnr, mKIAA1241 |
Location: |
Chr.13:100017994-100104073 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdp1 | NM_001081061.1 | 25 | 5220 | NP_001074530.1 |
| AK220262.1 | 15 | 3468 | BAD90187.1 | |
| BC144852.1 | 6 | 910 |
More Information
Set Membership: |
|

Genomic Map