Assay Details
Target Gene Details
Entrez Gene ID: | 544971 |
Gene Name: | B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
Gene Aliases: |
AI662220, AW049390, AW456745, B130055N23Rik, G630013P12Rik, TAF3B1, TFC5, TFIIIB150, TFIIIB90, Tfnr, mKIAA1241 |
Location: |
Chr.13:100017994-100104073 on Build GRCm38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdp1 | NM_001081061.1 | 38 | 7309 | NP_001074530.1 |
| AK078583.1 | 6 | 796 | ||
| AK090184.1 | 7 | 939 | BAC41127.1 | |
| AK220262.1 | 28 | 5557 | BAD90187.1 | |
| BC030359.1 | 10 | 1532 | AAH30359.1 | |
| BC144852.1 | 19 | 2999 |
More Information
Set Membership: |
|

Genomic Map