Assay Details
Target Gene Details
Entrez Gene ID: | 14062 |
Gene Name: | coagulation factor II (thrombin) receptor |
Gene Aliases: |
AI482343, Cf2r, Par1, ThrR |
Location: |
Chr.13:95601789-95618433 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2r | NM_010169.3 | 2 | 1203 | NP_034299.2 |
| AK085990.1 | 2 | 1244 | BAC39587.1 | |
| AK146194.1 | 2 | 1230 | BAE26968.1 | |
| AK159282.1 | 2 | 1258 | BAE34960.1 | |
| AK160032.1 | 2 | 1258 | BAE35575.1 | |
| BC031516.1 | 2 | 1203 | AAH31516.1 | |
| L03529.1 | 2 | 1158 | AAA40438.1 |
More Information
Set Membership: |
|

Genomic Map