Assay Details
Target Gene Details
Entrez Gene ID: | 14064 |
Gene Name: | coagulation factor II (thrombin) receptor-like 2 |
Gene Aliases: |
F730031A08, PAR3 |
Location: |
Chr.13:95696920-95702768 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2rl2 | NM_010170.4 | 2 | 958 | NP_034300.3 |
| AK089444.1 | 2 | 922 | BAC40886.1 | |
| AK135212.1 | 2 | 1025 | BAE22460.1 | |
| BC140985.1 | 2 | 913 | ||
| U92972.1 | 2 | 966 | AAC53137.1 |
Target Gene Details
Entrez Gene ID: | 544963 |
Gene Name: | IQ motif containing GTPase activating protein 2 |
Gene Aliases: |
4933417J23Rik, A630053O10, AI788777 |
Location: |
Chr.13:95627177-95891969 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqgap2 | NM_027711.1 | NP_081987.1 | ||
| AK142813.1 | BAE25199.1 | |||
| AK147360.1 | ||||
| BC052916.1 | AAH52916.1 | |||
| BC094939.1 | AAH94939.1 | |||
| BC150958.1 |
More Information
Set Membership: |
|

Genomic Map