Assay Details
Target Gene Details
Entrez Gene ID: | 271005 |
Gene Name: | kelch domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.12:69226917-69284646 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhdc1 | NM_178253.5 | 13 | 1261 | NP_839984.1 |
AK144621.1 | 13 | 1201 | BAE25974.1 | |
AY238938.1 | 13 | 1261 | AAO88963.1 | |
BC119567.2 | 13 | 1239 | ||
BC119568.2 | 13 | 1245 |
