Assay Details
Target Gene Details
Entrez Gene ID: | 208439 |
Gene Name: | kelch-like 29 |
Gene Aliases: |
A230106N14Rik, Gm68, Kbtbd9, mKIAA1921 |
Location: |
Chr.12:5077465-5375682 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl29 | NM_001164493.1 | 5 | 1588 | NP_001157965.1 |
| AK122572.1 | 5 | 1241 | BAC65854.1 | |
| BC138283.1 | 4 | 944 | ||
| BC145230.1 | 4 | 944 | ||
| BC145748.1 | 4 | 944 |
More Information
Set Membership: |
|

Genomic Map