Assay Details
Target Gene Details
Entrez Gene ID: | 22160 |
Gene Name: | twist basic helix-loop-helix transcription factor 1 |
Gene Aliases: |
AA960487, M-Twist, Pde, Ska10, Ska<m10Jus>, Twist, bHLHa38, pdt |
Location: |
Chr.12:33957671-33959831 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Twist1 | NM_011658.2 | 2 | 1076 | NP_035788.1 |
| BC033434.1 | 2 | 902 | AAH33434.1 | |
| BC083139.1 | 2 | 1076 | AAH83139.1 | |
| M63650.1 | 2 | 935 | AAA40515.1 |
More Information
Set Membership: |
|

Genomic Map