Assay Details
Target Gene Details
Entrez Gene ID: | 320581 |
Gene Name: | isopentenyl-diphosphate delta isomerase 2 |
Gene Aliases: |
4833405L16Rik, IPPI2 |
Location: |
Chr.13:8952863-8960935 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idi2 | NM_177197.4 | 5 | 1332 | NP_796171.1 |
| AK029364.1 | 5 | 1333 | BAC26418.1 | |
| AK081319.1 | 5 | 1333 | BAC38194.1 | |
| BC099586.1 | 5 | 1319 | AAH99586.1 | |
| BC125310.1 | 5 | 1318 | ||
| BC138100.1 | 5 | 1318 |
More Information
Set Membership: |
|

Genomic Map