Assay Details
Target Gene Details
Entrez Gene ID: | 18212 |
Gene Name: | neurotrophic tyrosine kinase, receptor, type 2 |
Gene Aliases: |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Location: |
Chr.13:58806569-59133970 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntrk2 | NM_001282961.1 | 1 | 116 | NP_001269890.1 |
| NM_008745.3 | 1 | 116 | NP_032771.1 | |
| AK018789.1 | BAB31412.1 | |||
| AK160789.1 | 1 | 117 | BAE36012.1 | |
| BY713295.1 | 1 | 111 |
More Information
Set Membership: |
|

Genomic Map