Assay Details
Target Gene Details
Entrez Gene ID: | 18212 |
Gene Name: | neurotrophic tyrosine kinase, receptor, type 2 |
Gene Aliases: |
GP145-TrkB/GP95-TrkB, Tkrb, trk-B, trkB |
Location: |
Chr.13:58806569-59133970 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntrk2 | NM_001025074.2 | 2 | 782 | NP_001020245.1 |
| NM_001282961.1 | 2 | 749 | NP_001269890.1 | |
| NM_008745.3 | 2 | 749 | NP_032771.1 | |
| AB377224.1 | 1 | 159 | ||
| AB377225.1 | 1 | 159 | ||
| AK009606.1 | 1 | 152 | ||
| AK018789.1 | 2 | 680 | BAB31412.1 | |
| AK028827.1 | 2 | 602 | ||
| AK147391.1 | 2 | 580 | BAE27880.1 | |
| AK147441.1 | 1 | 570 | BAE27914.1 | |
| AK160789.1 | 2 | 750 | BAE36012.1 | |
| BC052014.1 | 1 | 544 | AAH52014.2 | |
| M33385.1 | 1 | 121 | AAA40482.1 |
More Information
Set Membership: |
|

Genomic Map