Assay Details
Target Gene Details
Entrez Gene ID: | 12177 |
Gene Name: | BCL2/adenovirus E1B interacting protein 3-like |
Gene Aliases: |
C86132, D14Ertd719e, Nip3L, Nix |
Location: |
Chr.14:66985238-67008894 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bnip3l | NM_009761.3 | 6 | 2818 | NP_033891.1 |
AK004667.1 | 6 | 2765 | BAB23456.1 | |
AK155040.1 | 6 | 2789 | BAE33007.1 | |
AK168482.1 | 6 | 2819 | BAE40370.1 | |
AK169391.1 | 6 | 2819 | BAE41138.1 | |
AY057069.1 | 7 | 2839 | AAL18166.1 |
