Assay Details
Target Gene Details
Entrez Gene ID: | 76251 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 |
Gene Aliases: |
0610007P08Rik, 1700019D06Rik, 9330134C04Rik, Rad26l, Sr278 |
Location: |
Chr.13:63813415-63900302 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc6l2 | NM_001013608.2 | 19 | 4394 | NP_001013626.2 |
AJ250191.1 | 1 | 338 | CAC08498.1 | |
AK006112.1 | 3 | 808 | BAB24414.1 | |
BC026917.1 | 1 | 191 | AAH26917.1 | |
BC075679.1 | 4 | 1405 | AAH75679.1 |
