Assay Details
Target Gene Details
Entrez Gene ID: | 76251 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 6 like 2 |
Gene Aliases: |
0610007P08Rik, 1700019D06Rik, 9330134C04Rik, Rad26l, Sr278 |
Location: |
Chr.13:63813415-63900302 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc6l2 | NM_001013608.2 | 2 | 265 | NP_001013626.2 |
NM_023507.3 | 2 | 251 | NP_075996.2 | |
AF217319.1 | 1 | 68 | AAF73858.1 | |
AK002307.1 | 2 | 252 | BAB22002.1 | |
AK035054.1 | 2 | 217 | BAC28927.1 | |
AK049874.1 | 2 | 577 | ||
AK050437.1 | 1 | 101 | ||
AK082850.1 | BAC38652.1 | |||
BU148612.1 | 2 | 283 |
