Assay Details
Target Gene Details
Entrez Gene ID: | 75801 |
Gene Name: | RIKEN cDNA 4930447C04 gene |
Gene Aliases: |
4921504I02Rik, A930035O15Rik, Six6OS, Six6as, Six6os1 |
Location: |
Chr.12:72880983-72940083 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930447C04Rik | AY576418.1 |
Target Gene Details
Entrez Gene ID: | 20476 |
Gene Name: | sine oculis-related homeobox 6 |
Gene Aliases: |
Optx2, Six9 |
Location: |
Chr.12:72939945-72944899 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Six6 | NM_011384.4 | 1 | 119 | NP_035514.1 |
| AF050130.2 | 1 | 154 | AAC33850.1 | |
| AF135267.1 | 1 | 170 | AAD48911.1 | |
| AJ011787.1 | CAA09775.1 | |||
| AK017544.1 | 1 | 183 | ||
| AK029309.1 | 1 | 120 | BAC26387.1 | |
| BC138838.1 | 1 | 92 | ||
| BC138839.1 | 1 | 92 |
More Information
Set Membership: |
|

Genomic Map