Assay Details
Target Gene Details
Entrez Gene ID: | 12231 |
Gene Name: | butyrophilin, subfamily 1, member A1 |
Gene Aliases: |
Btn |
Location: |
Chr.13:23456984-23465914 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btn1a1 | NM_013483.3 | 3 | 420 | NP_038511.1 |
| AK021333.1 | 3 | 419 | BAB32377.1 | |
| AK142596.1 | 3 | 327 | BAE25122.1 | |
| AK145168.1 | 3 | 421 | BAE26272.1 | |
| BC009817.1 | 3 | 420 | AAH09817.1 | |
| BC011497.1 | 3 | 394 | AAH11497.1 | |
| BC031459.1 | 2 | 318 | AAH31459.1 | |
| S80642.1 | 1 | 143 | AAB35893.1 |
More Information
Set Membership: |
|

Genomic Map