Assay Details
Target Gene Details
Entrez Gene ID: | 50496 |
Gene Name: | E2F transcription factor 6 |
Gene Aliases: |
AI462434, E2F6b, EMA |
Location: |
Chr.12:16810903-16826752 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f6 | NM_033270.2 | 1 | 292 | NP_150373.2 |
| NR_027677.1 | 1 | 292 | ||
| AB221552.1 | ||||
| AF032131.1 | 1 | 270 | AAC53521.1 | |
| AK035116.1 | 1 | 289 | BAC28950.1 | |
| AK146792.1 | 1 | 288 | BAE27435.1 | |
| BC019166.1 | 1 | 288 | ||
| BC037656.1 | 1 | 169 | AAH37656.1 | |
| BC057929.1 | 1 | 280 | AAH57929.1 | |
| BY739130.1 | 1 | 293 | ||
| CJ075693.1 | 1 | 313 |
More Information
Set Membership: |
|

Genomic Map