Assay Details
Target Gene Details
Entrez Gene ID: | 68083 |
Gene Name: | PAK1 interacting protein 1 |
Gene Aliases: |
5830431I15Rik, 5930415H02Rik, AA419825, AI314040, AW556169, Gdpd1, PIP1 |
Location: |
Chr.13:41001010-41013033 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pak1ip1 | NM_026550.3 | 1 | 68 | NP_080826.2 |
| AF386076.1 | 1 | 55 | AAL40652.1 | |
| AK017959.1 | 1 | 55 | ||
| AK077846.1 | 1 | 56 | ||
| BY055321.1 | 1 | 70 | ||
| BY788898.1 | 1 | 54 |
More Information
Set Membership: |
|

Genomic Map