Assay Details
Target Gene Details
Entrez Gene ID: | 69048 |
Gene Name: | solute carrier family 30 (zinc transporter), member 5 |
Gene Aliases: |
1810010K08Rik, AF233321, ZNT5, ZTL1, ZnT-5, Zntl1 |
Location: |
Chr.13:100802645-100833563 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a5 | NM_022885.2 | 1 | 109 | NP_075023.2 |
| AF461761.1 | AAL96438.1 | |||
| AK051183.1 | 1 | 109 | BAC34549.1 | |
| AK088004.1 | BAC40089.1 | |||
| AK090176.1 | ||||
| BC033452.1 | AAH33452.1 |

Genomic Map