Assay Details
Target Gene Details
Entrez Gene ID: | 74338 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19 |
Gene Aliases: |
4632401C08Rik, B<0>AT1 |
Location: |
Chr.13:73679745-73700695 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a19 | NM_028878.3 | 1 | 180 | NP_083154.1 |
| AB096860.1 | 1 | 265 | BAC66209.1 | |
| AJ633679.1 | 1 | 189 | CAG17898.1 | |
| AK014544.1 | 1 | 181 | BAB29422.1 | |
| AK028784.1 | 1 | 181 | BAC26119.1 | |
| AK128923.1 | 1 | 180 | ||
| BC107411.1 | 1 | 181 | AAI07412.1 | |
| BC107412.2 | 1 | 185 | AAI07413.1 |
Target Gene Details
Entrez Gene ID: | 71390 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19, opposite strand |
Gene Aliases: |
5430425J12Rik |
Location: |
Chr.13:73699826-73709856 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc6a19os | NM_027780.1 | NP_082056.1 | ||
| AK017337.1 | BAB30695.1 |
More Information
Set Membership: |
|

Genomic Map